EMAIL THIS PAGE TO A FRIEND
Arquivos brasileiros de cardiologia
Human Tissue Kallikrein Activity in Angiographically Documented Chronic Stable Coronary Artery Disease.
PMID 26351984
Abstract
Human tissue kallikrein (hK1) is a key enzyme in the kallikrein-kinin system (KKS). hK1-specific amidase activity is reduced in urine samples from hypertensive and heart failure (HF) patients. The pathophysiologic role of hK1 in coronary artery disease (CAD) remains unclear. To evaluate hK1-specific amidase activity in the urine of CAD patientsMethods:Sixty-five individuals (18-75 years) who underwent cardiac catheterism (CATH) were included. Random midstream urine samples were collected immediately before CATH. Patients were classified in two groups according to the presence of coronary lesions: CAD (43 patients) and non-CAD (22 patients). hK1 amidase activity was estimated using the chromogenic substrate D-Val-Leu-Arg-Nan. Creatinine was determined using Jaffé's method. Urinary hK1-specific amidase activity was expressed as µM/(min · mg creatinine) to correct for differences in urine flow rates. Urinary hK1-specific amidase activity levels were similar between CAD [0.146 µM/(min ·mg creatinine)] and non-CAD [0.189 µM/(min . mg creatinine)] patients (p = 0.803) and remained similar to values previously reported for hypertensive patients [0.210 µM/(min . mg creatinine)] and HF patients [0.104 µM/(min . mg creatinine)]. CAD severity and hypertension were not observed to significantly affect urinary hK1-specific amidase activity. CAD patients had low levels of urinary hK1-specific amidase activity, suggesting that renal KKS activity may be reduced in patients with this disease.