EMAIL THIS PAGE TO A FRIEND
Molecular Medicine Reports
Poor permeability and absorption affect the activity of four alkaloids from Coptis.
PMID 26352530
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4759
Hydrochlorothiazide, crystalline
C7H8ClN3O4S2
H2910
Hydrochlorothiazide, meets USP testing specifications
C7H8ClN3O4S2