EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Endocrinology and Metabolism
Cytochrome c release from rat liver mitochondria is compromised by increased saturated cardiolipin species induced by sucrose feeding.
PMID 26353385