EMAIL THIS PAGE TO A FRIEND
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Triggering of Suicidal Erythrocyte Death by Ruxolitinib.
PMID 26356267
Abstract
The JAK1/JAK2 tyrosine kinase inhibitor ruxolitinib is widely used for the treatment of myeloproliferative neoplasm-associated myelofibrosis and other malignancies. Most important side effects include anemia. A common cause of anemia is accelerated suicidal death of erythrocytes or eryptosis, which is characterized by cell shrinkage and cell membrane scrambling with phosphatidylserine translocation to the erythrocyte surface. Mechanisms contributing to the triggering of eryptosis include oxidative stress, Ca2+ entry with increase of cytosolic Ca2+ activity ([Ca2+]i), and activation of distinct kinases, such as p38 mitogen activated protein (MAP) kinase. The present study explored whether and how ruxolitinib induces eryptosis. Phosphatidylserine exposure at the cell surface was estimated from annexin-V-binding, cell volume from forward scatter, hemolysis from hemoglobin release, [Ca2+]i from Fluo3-fluorescence, and ROS formation from DCFDA dependent fluorescence. A 48 hours exposure of human erythrocytes to ruxolitinib (25 µM) significantly increased the percentage of annexin-V-binding cells and significantly decreased forward scatter. Ruxolitinib did not significantly modify Fluo3-fluorescence and DCFDA fluorescence and the effect of ruxolitinib on annexin-V-binding was not significantly modified by removal of extracellular Ca2+. The effect of ruxolitinib on annexin-V-binding was, however, significantly blunted by the p38 MAP kinase inhibitor SB203580 and virtually abolished by the p38 MAP kinase inhibitor skepinone. Ruxolitinib triggers cell shrinkage and phospholipid scrambling of the erythrocyte cell membrane, an effect in part requiring p38 MAP kinase activity.