EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis of the alpha-Neu5Ac-(2----6)-Gal structure. Facile 6-O-sialylation following stannylene activation of an unprotected D-galactopyranoside.
PMID 2636042
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34980
Dibutyltin(IV) oxide, purum
C8H18OSn
183083
Dibutyltin(IV) oxide, 98%
C8H18OSn