PloS one
Nanoparticles Containing Curcumin Useful for Suppressing Macrophages In Vivo in Mice.
PMID 26361331
Abstract
To explore a novel method using liposomes to suppress macrophages, we screened food constituents through cell culture assays. Curcumin was one of the strongest compounds exhibiting suppressive effects on macrophages. We subsequently tried various methods to prepare liposomal curcumin, and eventually succeeded in preparing liposomes with sufficient amounts of curcumin to suppress macrophages by incorporating a complex of curcumin and bovine serum albumin. The diameter of the resultant nanoparticles, the liposomes containing curcumin, ranged from 60 to 100 nm. Flow cytometric analyses revealed that after intraperitoneal administration of the liposomes containing curcumin into mice, these were incorporated mainly by macrophages positive for F4/80, CD36, and CD11b antigens. Peritoneal cells prepared from mice injected in vivo with the liposomes containing curcumin apparently decreased interleukin-6-producing activities. Major changes in body weight and survival rates in the mice were not observed after administrating the liposomes containing curcumin. These results indicate that the liposomes containing curcumin are safe and useful for the selective suppression of macrophages in vivo in mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10798
Apigenin, ≥95.0% (HPLC)
C15H10O5
C8667
Cholesterol, Sigma Grade, ≥99%
C27H46O
C8503
Cholesterol, from sheep wool, ≥92.5% (GC), powder
C27H46O
C3045
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
C27H46O
H6167
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
C27H46O
C1231
Cholesterol, Plant-Derived, SyntheChol®, PharmaGrade, USP/NF, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C27H46O
C80105
Chrysin, 97%
C15H10O4
G0549
Corosolic acid from Lagerstroemia speciosa, ≥98% (HPLC)
C30H48O4
C7727
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
C21H20O6
C1386
Curcumin, from Curcuma longa (Turmeric), powder
C21H20O6
D7802
Daidzein, ≥98%, synthetic
C15H10O4
102032
Flavanone, 98%
C15H12O2
F2003
Flavone, ≥99.0%
C15H10O2
F3651
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
C21H11NO5S
46950
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
C21H11NO5S
F4274
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
C21H11NO5S
46951
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
C21H11NO5S
G6649
Genistein, synthetic, ≥98% (HPLC), powder
C15H10O5
G6776
Genistein, from Glycine max (soybean), ~98% (HPLC)
C15H10O5
Q4951
Quercetin, ≥95% (HPLC), solid
C15H10O7
R5010
Resveratrol, ≥99% (HPLC)
C14H12O3
S5442
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
C27H46O
U6753
Ursolic acid, ≥90%
C30H48O3