EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Annexin A10 expression in colorectal cancers with emphasis on the serrated neoplasia pathway.
PMID 26361422
Abstract
To validate the utility of Annexin A10 as a surrogate marker of the serrated neoplasia pathway in invasive colorectal cancers (CRCs). A total of 1133 primary CRC patients who underwent surgical resection at Seoul National University Hospital between January 2004 and December 2007 were enrolled. Expression of Annexin A10 was evaluated by immunohistochemistry using tissue microarray and paired to our findings on clinicopathologic and molecular characteristics of each individual. CpG island methylator phenotype was determined by MethyLight assay and microsatellite instability was determined by high performance liquid chromatography. KRAS and BRAF mutation status was evaluated by direct sequencing and allele-specific PCR. Univariate and stage-specific survival analyses were performed to reveal the prognostic value of Annexin A10 expression. Annexin A10 expression was observed in 66 (5.8%) of the 1133 patients. Annexin A10 expression was more commonly found in females and was associated with proximal location, ulcerative gross type, advanced T category, N category and TNM stage. CRCs with Annexin A10 expression showed an absence of luminal necrosis, luminal serration and mucin production. CRCs with Annexin A10 expression were associated with CpG island methylator phenotype, microsatellite instability and BRAF mutation. In survival analysis, Annexin A10 expression was associated with poor overall survival and progression-free survival, especially in stage IV CRCs. Annexin A10 expression is associated with poor clinical behavior and can be used a supportive surrogate marker of the serrated neoplasia pathway in invasive CRCs.