EMAIL THIS PAGE TO A FRIEND
American journal of surgery
Defining surgical outcomes and quality of life in massive ventral hernia repair: an international multicenter prospective study.
PMID 26362202
Abstract
Our goal was to set criteria for massive ventral hernia and to compare surgical outcomes and quality of life after ventral hernia repair (VHR). The International Hernia Mesh Registry was queried for patients undergoing VHR from 2007 to 2013. Defect was categorized as massive if the width or length was greater than 15 cm or area greater than 150 cm(2). Massive VHR was compared to regular VHR. A total of 878 patients underwent VHR: 436 open, 442 laparoscopic with 13 deaths (1.5%) and 45 hernia recurrences (5.1%). Of those, 158 patients (18%) met criteria for massive VHR. Massive VHR patients had longer length of stay (LOS) and operative time and more hematomas, wound infections, wound complications, and pneumonias (P < .05). On multivariate analysis, LOS was longer, and early postoperative pain and activity limitation were greater in massive VHRs (P < .01). Massive VHR in the laparoscopic approach resulted in greater long-term mesh sensation (P < .01). VHR in massive hernias have increased rates of complications and longer LOS.