EMAIL THIS PAGE TO A FRIEND
Human pathology
Diagnostic value of maspin in distinguishing adenocarcinoma from benign biliary epithelium on endoscopic bile duct biopsy.
PMID 26362203
Abstract
Histopathologic distinction between benign and malignant epithelia on endoscopic bile duct biopsy can be extremely challenging due to small sample size, crush artifact, and a propensity for marked inflammatory and reactive changes after stent placement. Our previous studies have shown that the insulin-like growth factor II mRNA-binding protein 3, S100P, and the von Hippel-Lindau gene product (pVHL) can help the distinction. This study analyzed 134 endoscopic bile duct biopsy specimens (adenocarcinoma 45, atypical 31, and benign 58) by immunohistochemistry for the expression of maspin, a serine protease inhibitor. The results demonstrated that (1) maspin expression was more frequently detected in malignant than in benign biopsies; (2) malignant biopsies frequently showed diffuse, strong/intermediate, and combined nuclear/cytoplasmic staining patterns for maspin, which were much less commonly seen in benign biopsies; (3) the malignant staining patterns for maspin observed in atypical biopsies were consistent with follow-up data showing that 67% of these patients were subsequently diagnosed with adenocarcinoma; (4) a maspin+/S100P+/pVHL- staining profile was seen in 75% of malignant biopsies but in none of the benign cases. These observations demonstrate that maspin is a useful addition to the diagnostic immunohistochemical panel (S100P, pVHL, and insulin-like growth factor II mRNA-binding protein 3) to help distinguish malignant from benign epithelia on challenging bile duct biopsies.