EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Genome-wide RNA sequencing analysis of quorum sensing-controlled regulons in the plant-associated Burkholderia glumae PG1 strain.
PMID 26362987