EMAIL THIS PAGE TO A FRIEND
Agressologie: revue internationale de physio-biologie et de pharmacologie appliquees aux effets de l'agression
Further clinical experiences with optical isomers of the L and D clorothepin.
PMID 263672
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O111
Octoclothepin maleate salt, solid
C19H21ClN2S · C4H4O4