EMAIL THIS PAGE TO A FRIEND
Food additives & contaminants. Part B, Surveillance
Natural incidence of zearalenone in Croatian pig feed, urine and meat in 2014.
PMID 26367461
Abstract
The aim of this study was to determine the levels of zearalenone (ZEN) in different feed materials and feedstuffs for pigs, as well as in pig urine and pig meat following contaminated feed consumption. In total, 253 feed material and feedstuff samples were collected from Croatian pig farms. The results revealed the presence of ZEN in significant concentrations, the maximal being found in maize (5522 µg/kg), wheat (3366 µg/kg) and pig fattening feed (1949 µg/kg). In farms in which high feed contamination and pig hyperestrogenism were observed, samples of pig urine (n=30) and meat (n=30) were retrieved as well. The mean ZEN concentrations in pig urine and pig meat were 206±20.6 µg/L and 0.62±0.14 µg/kg, respectively. Despite high contamination of feedstuffs responsible for farmed pigs' intoxication, ZEN levels determined in pig meat were shown to be of little significance for human safety.