EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Sciences
A New HPLC-MS Method for Measuring Maslinic Acid and Oleanolic Acid in HT29 and HepG2 Human Cancer Cells.
PMID 26370984
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
HT29 Cell Line human, Caucasian colon adenocarcinoma grade II