EMAIL THIS PAGE TO A FRIEND
Applied Biochemistry and Biotechnology
Production and Characterization of Surfactin-Type Lipopeptides as Bioemulsifiers Produced by a Pinctada martensii-Derived Bacillus mojavensis B0621A.
PMID 26373943