EMAIL THIS PAGE TO A FRIEND
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment
Development and validation of an UHPLC-MS/MS method for the determination of mycotoxins in grass silages.
PMID 26374621