EMAIL THIS PAGE TO A FRIEND
International Journal of Pharmaceutics
Artemisinin nanoformulation suitable for intravenous injection: Preparation, characterization and antimalarial activities.
PMID 26383839
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
361593
Artemisinin, 98%
C15H22O5