EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Insights into the strong in-vitro anticancer effects for bis(triphenylphosphane)iminium compounds having perchlorate, tetrafluoridoborate and bis(chlorido)argentate anions.
PMID 26384162
Abstract
Three new compounds containing the bis(triphenylphosphane)iminium cation (PPN(+)) with ClO4(-), BF4(-) and [AgCl2](-) as counter anions have been synthesized and structurally characterized. The two derivatives with ClO4(-) and BF4(-) were found to be isostructural by single crystal X-ray diffraction. Interestingly, the three compounds show extremely potent antiproliferative effects against the human cancer cell line SKOV3. To gain insights into the possible mechanisms of biological action, several intracellular targets have been considered. Thus, DNA binding has been evaluated, as well as the effects of the compounds on the mitochondrial function. Furthermore, the compounds have been tested as possible inhibitors of the seleno-enzyme thioredoxin reductase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5428
Tetramethylrhodamine methyl ester perchlorate, ≥95%
C25H25ClN2O7