EMAIL THIS PAGE TO A FRIEND
Journal of biotechnology
Secretory production of antimicrobial peptides in Escherichia coli using the catalytic domain of a cellulase as fusion partner.
PMID 26387445
Abstract
Antimicrobial peptides (AMPs) are small molecules which serve as essential components of the innate immune system in various organisms. AMPs possess a broad spectrum of antimicrobial activities. However, the scaled production of such peptides in Escherichia coli faces many difficulties because of their small size and toxicity to the host. Here, we described a new fusion strategy to extracellularly produce significant amounts of these antimicrobial peptides in recombinant E. coli at significant amount. Employing the catalytic domain of a cellulase (Cel-CD) from Bacillus subtilis KSM-64 as the fusion partner, five recombinant antimicrobial peptides were confirmed to accumulate in the culture medium at concentrations ranging from 184 mg/L to 297 mg/L. The radical diffusion experiment demonstrated that the released model antimicrobial peptide, bombinin, had antibacterial activities against both E. coli and Staphylococcus aureus. This strategy will be suitable for the production of antimicrobial peptides and other toxicity proteins.