EMAIL THIS PAGE TO A FRIEND
Toxicology and Applied Pharmacology
Estimation of tetrabromobisphenol A (TBBPA) percutaneous uptake in humans using the parallelogram method.
PMID 26387765
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
330396
3,3′,5,5′-Tetrabromobisphenol A, 97%
C15H12Br4O2