EMAIL THIS PAGE TO A FRIEND
Laboratory Investigation; a Journal of Technical Methods and Pathology
Basal cells of the human airways acquire mesenchymal traits in idiopathic pulmonary fibrosis and in culture.
PMID 26390052