EMAIL THIS PAGE TO A FRIEND
ACS Nano
Cuboplexes: Topologically Active siRNA Delivery.
PMID 26390340
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323616
3′-Deoxythymidine, 98%
C10H14N2O4