EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Structural and Biophysical Characterization of the Interactions between Calmodulin and the Pleckstrin Homology Domain of Akt.
PMID 26391397
Abstract
The translocation of Akt, a serine/threonine kinase, to the plasma membrane is a critical step in the Akt activation pathway. It is established that membrane binding of Akt is mediated by direct interactions between its pleckstrin homology domain (PHD) and phosphatidylinositol 3,4,5-trisphosphate (PI(3,4,5)P3). There is now evidence that Akt activation in many breast cancer cells is also modulated by the calcium-binding protein, calmodulin (CaM). Upon EGF stimulation of breast cancer cells, CaM co-localizes with Akt at the plasma membrane to enhance activation. However, the molecular details of Akt(PHD) interaction with CaM are not known. In this study, we employed NMR, biochemical, and biophysical techniques to characterize CaM binding to Akt(PHD). Our data show that CaM forms a tight complex with the PHD of Akt (dissociation constant = 100 nm). The interaction between CaM and Akt(PHD) is enthalpically driven, and the affinity is greatly dependent on salt concentration, indicating that electrostatic interactions are important for binding. The CaM-binding interface in Akt(PHD) was mapped to two loops adjacent to the PI(3,4,5)P3 binding site, which represents a rare CaM-binding motif and suggests a synergistic relationship between CaM and PI(3,4,5)P3 upon Akt activation. Elucidation of the mechanism by which Akt interacts with CaM will help in understanding the activation mechanism, which may provide insights for new potential targets to control the pathophysiological processes of cell survival.