EMAIL THIS PAGE TO A FRIEND
Biomedical Materials (Bristol, England)
Novel cerium doped glass-reinforced hydroxyapatite with antibacterial and osteoconductive properties for bone tissue regeneration.
PMID 26391473
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
450146
Calcium hydroxide, 99.995% trace metals basis
H2CaO2
C7263
Calcium phosphate dibasic, 98.0-105.0%
HCaO4P
21177
Calcium phosphate dibasic, puriss. p.a., anhydrous, >98.5% (KT)
HCaO4P
C8606
Dibasic Calcium Phosphate, meets USP testing specifications
HCaO4P
P5811
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
H3O4P
466123
Phosphoric acid, crystalline, ≥99.999% trace metals basis
H3O4P
452289
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
H3O4P
345245
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
H3O4P
79617
Phosphoric acid, BioUltra, ≥85% (T)
H3O4P
04102
Phosphoric acid, puriss., ≥99%
H3O4P
40278
Phosphoric acid, semiconductor grade MOS PURANAL (Honeywell 17938), ≥85%
H3O4P
40266
Phosphoric acid, semiconductor grade PURANAL (Honeywell 17861), ≥85%
H3O4P
612162
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
H3O4P
699608
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
H3O4P
698822
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
H3O4P
W290017
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
H3O4P
609781
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
H316O4P
431419
Phosphorus pentoxide, ≥99.99% trace metals basis
O5P2
79609
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
O5P2
79610
Phosphorus pentoxide, desiccant, with moisture indicator
O5P2
298220
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
O5P2
214701
Phosphorus pentoxide, ReagentPlus®, 99%
O5P2
P4864
Propidium iodide solution, solution (1.0 mg/ml in water)
C27H34I2N4
330361
Soda ash, (99% as sodium carbonate or 58% as sodium oxide), technical grade, light
CNa2O3
791768
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
S7795
Sodium carbonate, BioXtra, ≥99.0%
CNa2O3
204420
Sodium carbonate, 99.995% trace metals basis
CNa2O3
451614
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
CNa2O3
71345
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
CNa2O3
28-2170
Sodium carbonate, JIS special grade, ≥99.5%
CNa2O3