EMAIL THIS PAGE TO A FRIEND
Acta Biomaterialia
Co-immobilization of semaphorin3A and nerve growth factor to guide and pattern axons.
PMID 26391495
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D13208
1,4-Diaminobutane, 99%
C4H12N2
294993
Ammonia, anhydrous, ≥99.98%
H3N
779423
Ammonia solution, 4 M in methanol
H3N
718939
Ammonia solution, 0.4 M in THF
H3N
608777
Ammonia-14N, 99.99 atom % 14N
H314N
M9768
Cysteamine, ~95%
C2H7NS
30070
Cysteamine, ≥98.0% (RT)
C2H7NS
480975
Hydroxylamine-O-sulfonic acid, 99.999%
H3NO4S
213136
Hydroxylamine-O-sulfonic acid, 97%
H3NO4S
55493
Hydroxylamine-O-sulfonic acid, purum, ≥95.0% (RT)
H3NO4S
227978
Hydroxylamine-O-sulfonic acid, technical grade, 90%
H3NO4S
G5792
L-Glutamine, PharmaGrade, Ajinomoto, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H10N2O3
W368401
L-Glutamine
C5H10N2O3
W262706
Levulinic acid, ≥97%, FG
C5H8O3
L2009
Levulinic acid, 98%
C5H8O3
W262701
Levulinic acid, natural, 99%, FG
C5H8O3
P0130
Progesterone, ≥99%
C21H30O2
P6149
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
C21H30O2
P8783
Progesterone, powder, BioReagent, suitable for cell culture
C21H30O2
P3972
Progesterone, meets USP testing specifications
C21H30O2
792519
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
CHNaO3
S5761
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
CHNaO3
S6297
Sodium bicarbonate, BioXtra, 99.5-100.5%
CHNaO3
S4019
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
CHNaO3
487031
Sodium bicarbonate-12C, 99.9 atom % 12C
12CHNaO3
S9133
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Na2O3Se
S5261
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Na2O3Se
214485
Sodium selenite, 99%
Na2O3Se
T0886
Triethylamine, ≥99%
C6H15N
90335
Triethylamine, BioUltra, ≥99.5% (GC)
C6H15N