EMAIL THIS PAGE TO A FRIEND
Bioprocess and Biosystems Engineering
Optimization of diacylglycerol production by glycerolysis of fish oil catalyzed by Lipozyme TL IM with Tween 65.
PMID 26391508
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7765
1-Oleoyl-rac-glycerol, ≥99%
C21H40O4
49960
1-Oleoyl-rac-glycerol, technical, ~40% (TLC)
C21H40O4