EMAIL THIS PAGE TO A FRIEND
Chirality
Unequivocal Enantiomeric Identification and Analysis of 10 Chiral Pesticides in Fruit and Vegetables by QuEChERS Method Combined With Liquid Chromatography-Quadruple/Linear Ion Trap Mass Spectrometry Determination.
PMID 26392120
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N12380
Metalaxyl, analytical standard
C15H21NO4
442818
Triadimefon, analytical standard
C14H16ClN3O2