EMAIL THIS PAGE TO A FRIEND
Biochimica et Biophysica Acta
Functional characterization of a fatty acid double-bond hydratase from Lactobacillus plantarum and its interaction with biosynthetic membranes.
PMID 26392225