EMAIL THIS PAGE TO A FRIEND
Biochemistry and cell biology = Biochimie et biologie cellulaire
Verification and refinement of cellular glycosphingolipid profiles using HPLC.
PMID 26393781
Abstract
Glycosphingolipids (GSLs) are hybrid molecules consisting of the sphingolipid ceramide linked to a mono- or oligo-saccharide. In comparison to other membrane lipids, the family of GSLs stands out because of the extensive variation in the carbohydrate headgroup. GSLs are cell surface binding partners, in cis with growth factor receptors, and in trans with bacterial toxins and viruses, and are among the host-derived membrane components of viral particles, including those of HIV. In spite of their biological relevance, GSL profiles of commonly used cell lines have been analyzed to different degrees. Here, we directly compare the GSL complements from CHO-K1, COS-7, HeLa, HEK-293, HEPG2, Jurkat, and SH-SY5Y cells using an HPLC-based method requiring modest amounts of material. Compared to previous studies, the HPLC-based analyses provided more detailed information on the complexity of the cellular GSL complement, qualitatively as well as quantitatively. In particular for cells expressing multiple GSLs, we found higher numbers of GSL species, and different levels of abundance. Our study thus extends our knowledge of biologically relevant lipids in widely used cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7753
Maltoheptaose, ≥60% (HPLC)
C42H72O36