EMAIL THIS PAGE TO A FRIEND
Molecular Pharmaceutics
Antibody-Drug Conjugates (ADCs) Derived from Interchain Cysteine Cross-Linking Demonstrate Improved Homogeneity and Other Pharmacological Properties over Conventional Heterogeneous ADCs.
PMID 26393951