EMAIL THIS PAGE TO A FRIEND
Immunologic Research
IL-10 restricts dendritic cell (DC) growth at the monocyte-to-monocyte-derived DC interface by disrupting anti-apoptotic and cytoprotective autophagic molecular machinery.
PMID 26395023