EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Selective quantitation of the neurotoxin BMAA by use of hydrophilic-interaction liquid chromatography-differential mobility spectrometry-tandem mass spectrometry (HILIC-DMS-MS/MS).
PMID 26396078
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS000144
[(2-Aminoethyl)amino]acetic acid, AldrichCPR
C4H10N2O2
D3758
DL-2,4-Diaminobutyric acid dihydrochloride
C4H10N2O2 · 2HCl