EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Routes of Ca2+ Shuttling during Ca2+ Oscillations: FOCUS ON THE ROLE OF MITOCHONDRIAL Ca2+ HANDLING AND CYTOSOLIC Ca2+ BUFFERS.
PMID 26396196