EMAIL THIS PAGE TO A FRIEND
Molecular Medicine Reports
Azorella compacta methanolic extract induces apoptosis via activation of mitogen-activated protein kinase.
PMID 26397193
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0466
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin, ≥90% (HPLC), powder
C30H34F3N5O13