EMAIL THIS PAGE TO A FRIEND
Hormone Research in p?diatrics
Concurrent Confirmation and Differential Diagnosis of Congenital Adrenal Hyperplasia from Dried Blood Spots: Application of a Second-Tier LC-MS/MS Assay in a Cross-Border Cooperation for Newborn Screening.
PMID 26397944