EMAIL THIS PAGE TO A FRIEND
Cells Tissues Organs
Repulsive Guidance Molecules a, b and c Are Skeletal Muscle Proteins, and Repulsive Guidance Molecule a Promotes Cellular Hypertrophy and Is Necessary for Myotube Fusion.
PMID 26397945