EMAIL THIS PAGE TO A FRIEND
Biomaterials
Patient-specific cardiovascular progenitor cells derived from integration-free induced pluripotent stem cells for vascular tissue regeneration.
PMID 26398309
Abstract
Tissue-engineered blood vessels (TEBVs) are promising in regenerating a live vascular replacement. However, the vascular cell source is limited, and it is crucial to develop a scaffold that accommodates new type of vascular progenitor cells and facilitates in vivo lineage specification of the cells into functional vascular smooth muscle cells (VSMCs) to regenerate vascular tissue. In the present study, integration-free human induced pluripotent stem cells (hiPSCs) were established from patient peripheral blood mononuclear cells through episomal vector nucleofection of reprogramming factors. The established hiPSCs were then induced into mesoderm-originated cardiovascular progenitor cells (CVPCs) with a highly efficient directed lineage specification method. The derived CVPCs were demonstrated to be able to differentiate into functional VSMCs. Subcutaneous implantation of CVPCs seeded on macroporous nanofibrous poly(l-lactide) scaffolds led to in vivo VSMC lineage specification and matrix deposition inside the scaffolds. In summary, we established integration-free patient-specific hiPSCs from peripheral blood mononuclear cells, derived CVPCs through directed lineage specification, and developed an advanced scaffold for these progenitor cells to further differentiate in vivo into VSMCs and regenerate vascular tissue in a subcutaneous implantation model. This study has established an efficient patient-specific approach towards in vivo regeneration of vascular tissue.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56454
1-Thioglycerol, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C3H8O2S
M1753
1-Thioglycerol, ≥97%
C3H8O2S
M6145
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
C3H8O2S
M2172
1-Thioglycerol, BioXtra, ≥97%
C3H8O2S
88640
1-Thioglycerol, ≥99.0% (GC)
C3H8O2S
A5960
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
C6H8O6
A7506
L-Ascorbic acid, reagent grade, crystalline
C6H8O6
A0278
L-Ascorbic acid, reagent grade
C6H8O6
A4403
L-Ascorbic acid, powder, cell culture tested, γ-irradiated
C6H8O6
A4544
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
C6H8O6
A2218
L-Ascorbic acid, meets USP testing specifications
C6H8O6
A2174
L-Ascorbic acid, plant cell culture tested
C6H8O6
A2343
L-Ascorbic acid, meets USP testing specifications
C6H8O6
95209
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
C6H8O6
33034
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.7-100.5% (oxidimetric)
C6H8O6
95210
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
C6H8O6
47863
L-Ascorbic acid, analytical standard
C6H8O6
255564
L-Ascorbic acid, ACS reagent, ≥99%
C6H8O6
A92902
L-Ascorbic acid, 99%
C6H8O6
W210901
L-Ascorbic acid, FCC, FG
C6H8O6
V-011
Retinol solution, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, certified reference material
C20H30O