EMAIL THIS PAGE TO A FRIEND
Journal of Alzheimer's disease : JAD
Prolonged In Vivo Retention of a Cathepsin D Targeted Optical Contrast Agent in a Mouse Model of Alzheimer's Disease.
PMID 26401930
Abstract
Cathepsin D (CatD) is a lysosomal protease that is elevated early in Alzheimer's disease (AD). We have previously developed a Targeted contrast agent (CA) to detect CatD activity in vivo, consisting of a magnetic resonance imaging/fluorescent moiety linked to a cell penetrating peptide (CPP) by means of a CatD cleavage site and have demonstrated its uptake in the brain of an AD mouse model. The purpose of this study was to characterize the in vivo retention of a near infra-red fluorescent dye labeled version of this CA. Six adult C57Bl/6 wild-type mice and six adult 5XFAD transgenic AD mice were studied using a small animal imaging system at five and twelve months of age using our novel Targeted CA, or two different control CAs; a Non-Targeted (lacking the CatD cleavage site) and a Non-Penetrating (lacking the CPP). Following intravenous CA administration, the optical signal was recorded within the brain and uptake and washout curves were measured and fitted to a one-phase exponential decay curve. In all wild-type and 5XFAD mice, the washout of the Targeted CA that included a CPP domain was significantly slower than the washout of the Non-Penetrating and Non-Targeted CA. Furthermore, the washout of the CatD Targeted CA was significantly slower in the 5XFAD mice compared to the age matched wild-type controls (p <  0.05) at 5 and 12 months of age. Control CAs showed no differences in washout. The prolonged retention of the CatD targeted CA in 5XFAD mice suggests this agent may be useful for AD detection.