EMAIL THIS PAGE TO A FRIEND
Faraday Discussions
Thioether-triphenolate bimetallic iron(iii) complexes as robust and highly efficient catalysts for cycloaddition of carbon dioxide to epoxides.
PMID 26402776