EMAIL THIS PAGE TO A FRIEND
Journal of Pharmacological Sciences
Retinal region-dependent susceptibility of capillaries to high-concentration oxygen exposure and vascular endothelial growthi??A?factor receptor inhibition in neonatal mice.
PMID 26403253