EMAIL THIS PAGE TO A FRIEND
Hepatology (Baltimore, Md.)
Activation of the p62-Keap1-NRF2 pathway protects against ferroptosis in hepatocellular carcinoma cells.
PMID 26403645
Abstract
Ferroptosis is a recently recognized form of regulated cell death caused by an iron-dependent accumulation of lipid reactive oxygen species. However, the molecular mechanisms regulating ferroptosis remain obscure. Here, we report that nuclear factor erythroid 2-related factor 2 (NRF2) plays a central role in protecting hepatocellular carcinoma (HCC) cells against ferroptosis. Upon exposure to ferroptosis-inducing compounds (e.g., erastin, sorafenib, and buthionine sulfoximine), p62 expression prevented NRF2 degradation and enhanced subsequent NRF2 nuclear accumulation through inactivation of Kelch-like ECH-associated protein 1. Additionally, nuclear NRF2 interacted with transcriptional coactivator small v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog proteins such as MafG and then activated transcription of quinone oxidoreductase-1, heme oxygenase-1, and ferritin heavy chain-1. Knockdown of p62, quinone oxidoreductase-1, heme oxygenase-1, and ferritin heavy chain-1 by RNA interference in HCC cells promoted ferroptosis in response to erastin and sorafenib. Furthermore, genetic or pharmacologic inhibition of NRF2 expression/activity in HCC cells increased the anticancer activity of erastin and sorafenib in vitro and in tumor xenograft models. These findings demonstrate novel molecular mechanisms and signaling pathways of ferroptosis; the status of NRF2 is a key factor that determines the therapeutic response to ferroptosis-targeted therapies in HCC cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7781
Erastin, ≥98% (HPLC)
C30H31ClN4O4
SHCLNG MISSION® shRNA, Bacterial Stock