EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Sciences
Use of Copper to Selectively Inhibit Brachionus calyciflorus (Predator) Growth in Chlorella kessleri (Prey) Mass Cultures for Algae Biodiesel Production.
PMID 26404247