EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
BTLA marks a less-differentiated tumor-infiltrating lymphocyte subset in melanoma with enhanced survival properties.
PMID 26405566
Abstract
In a recent adoptive cell therapy (ACT) clinical trial using autologous tumor-infiltrating lymphocytes (TILs) in patients with metastatic melanoma, we found an association between CD8