EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
Antitumor activity of epigenetic immunomodulation combined with CTLA-4 blockade in syngeneic mouse models.
PMID 26405573
Abstract
The multifaceted immunomodulatory activity of DNA hypomethylating agents improves immunogenicity and immune recognition of neoplastic cells; thus, we predicted they could be utilized to design new immunotherapeutic combinations in cancer. Testing this hypothesis, the antitumor efficacy of the DNA hypomethylating agent 5-aza-2'-deoxycytidine (5-AZA-CdR) combined with the anti-CTLA-4 monoclonal antibody (mAb) 9H10 in syngeneic transplantable murine models was investigated. Murine mammary carcinoma TS/A or mesothelioma AB1 cells were injected in BALB/c, athymic nude, and SCID/Beige mice that were treated with 5-AZA-CdR, mAb 9H10, or their combination. Tumor volumes were captured at different time-points; molecular and immunohistochemical assays investigated changes in neoplastic and normal tissues. A significant antitumor effect of 5-AZA-CdR combined with mAb 9H10 was found: compared to controls, a 77% (