EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
Survivin-targeted immunotherapy drives robust polyfunctional T cell generation and differentiation in advanced ovarian cancer patients.
PMID 26405584
Abstract
DepoVax™ is an innovative and strongly immunogenic vaccine platform. Survivin is highly expressed in many tumor types and has reported prognostic value. To generate tumor-specific immune response, a novel cancer vaccine was formulated in DepoVax platform (DPX-Survivac) using survivin HLA class I peptides. Safety and immune potency of DPX-Survivac was tested in combination with immune-modulator metronomic cyclophosphamide in ovarian cancer patients. All the patients receiving the therapy produced antigen-specific immune responses; higher dose vaccine and cyclophosphamide treatment generating significantly higher magnitude responses. Strong T cell responses were associated with differentiation of naïve T cells into central/effector memory (CM/EM) and late differentiated (LD) polyfunctional antigen-specific CD4(+) and CD8(+) T cells. This approach enabled rapid de novo activation/expansion of vaccine antigen-specific CD8(+) T cells and provided a strong rationale for further testing to determine clinical benefits associated with this immune activation. These data represent vaccine-induced T cell activation in a clinical setting to a self-tumor antigen previously described only in animal models.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6250
2-Mercaptoethanol, ≥99.0%
C2H6OS
M3148
2-Mercaptoethanol, for molecular biology, for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
C2H6OS
63689
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
C2H6OS
I9516
2-Propanol, BioReagent, for molecular biology, ≥99.5%
C3H8O
278475
2-Propanol, anhydrous, 99.5%
C3H8O
675431
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
C3H8O
437522
2-Propanol, ACS reagent, ≥99.5%
C3H8O
59304
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C3H8O
40301
2-Propanol, semiconductor grade VLSI PURANAL (Honeywell 17604)
C3H8O
40219
2-Propanol, semiconductor grade PURANAL (Honeywell 17829)
C3H8O
270490
2-Propanol, 99.5%, HPLC grade
C3H8O
15-2360
2-Propanol, for HPLC
C3H8O
733458
2-Propanol, electronic grade, 99.999% trace metals basis
C3H8O
A6926
3-Amino-9-ethylcarbazole, tablet
C14H14N2
A5754
3-Amino-9-ethylcarbazole, ≥95%, powder
C14H14N2
W292907
Isopropyl alcohol, ≥99.7%, FCC, FG
C3H8O
G3126
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
C5H10N2O3
G5792
L-Glutamine, PharmaGrade, Ajinomoto, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H10N2O3
G8540
L-Glutamine, meets USP testing specifications, cell culture tested, 99.0-101.0%, from non-animal source
C5H10N2O3
G6392
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
C5H10N2O3
W368401
L-Glutamine
C5H10N2O3
79346
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
C36H56O8
P8139
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
C36H56O8