EMAIL THIS PAGE TO A FRIEND
Journal of Biomaterials Science. Polymer Edition
Functionalization of electrospun poly(caprolactone) fibers for pH-controlled delivery of doxorubicin hydrochloride.
PMID 26406285