EMAIL THIS PAGE TO A FRIEND
Optics express
Measuring nematic liquid crystal anchoring energy using whispering gallery modes.
PMID 26406690
Abstract
Whispering gallery modes (WGMs) around the liquid crystal droplets are sensitive to the director orientation at the interface. An electric field was applied for changing the director orientation of a nematic liquid crystal droplet surrounded by polydimethylsiloxane (PDMS), which induced vertical director orientation at the interface. WGMs were shifted by varying the electric field intensity, and WGM shift-dependent director response was calculated. From this calculation we obtained the extrapolation length of 0.35 ± 0.02 μm. These results indicate that WGM sensitivity can be used for obtaining the surface properties of liquid crystals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R6626
Rhodamine B, ≥95% (HPLC)
C28H31ClN2O3
83689
Rhodamine B, for fluorescence
C28H31ClN2O3