EMAIL THIS PAGE TO A FRIEND
Ecotoxicology and Environmental Safety
The concentration, source and potential human health risk of heavy metals in the commonly consumed foods in Bangladesh.
PMID 26406870