EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Interleukin 10 (IL-10)-mediated Immunosuppression: MARCH-I INDUCTION REGULATES ANTIGEN PRESENTATION BY MACROPHAGES BUT NOT DENDRITIC CELLS.
PMID 26408197