EMAIL THIS PAGE TO A FRIEND
Journal of Chemical Neuroanatomy
Imaging the time-integrated cerebral metabolic activity with subcellular resolution through nanometer-scale detection of biosynthetic products deriving from (13)C-glucose.
PMID 26409162