EMAIL THIS PAGE TO A FRIEND
Journal of Dairy Science
The effect of grape seed and grape marc meal extract on milk performance and the expression of genes of endoplasmic reticulum stress and inflammation in the liver of dairy cows in early lactation.
PMID 26409958